Would you like to log in again?
CMRL 1066 Medium Mammalian Cell Culture Medium, HiMedia
Supplier: HiMedia Laboratories
AT110-20L AT110-1L AT110-5L AT110-5L AT110-1L AT110-20L
97065-316EA 515.21 USD InStock
CA97065-316 97065-316 CA97065-314 CA97065-312 97065-314 97065-312
CMRL 1066 Medium Mammalian Cell Culture Medium, HiMedia
Cell Culture Media
CMRL media are based on the composition of Medium 199.
  • Based on the composition of Medium 199
  • Chemically defined medium formulated for long term cultivation of mouse L cells
  • Supports growth of a variety of cell lines in the presence of serum
Nucleic acid derivatives, coenzymes omitted in H597 modification were retained in CMRL 1066. CMRL 1066 is a chemically defined medium formulated for long term cultivation of mouse L cells in the absence of protein supplement. Although originally designed for use in serum free conditions, this medium supports growth of a variety of cell lines in the presence of serum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty