Would you like to log in again?
Aufricht Retractor, OR Grade, Sklar®
Supplier: Sklar
41-1410
10196-670EA 295.94 USD InStock
10196-670
Aufricht Retractor, OR Grade, Sklar®
Retractors
Sklar's® Aufricht Nasal Retractor is an ENT instrument commonly used for holding back the nostrils in order to provide better visulaization of the nasal cavity
This product features an 11mm x 45mm fenestrated blade that angles downward, and has an overall length of 61/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty