Would you like to log in again?
AAV Rep-Cap Vectors, Cell Biolabs
Supplier: Cell Biolabs
VPK-424 VPK-426 VPK-423 VPK-425 VPK-421 VPK-422
102966-360EA 683.08 USD InStock
102966-360 102965-558 102965-556 102966-358 102966-362 102966-364
AAV Rep-Cap Vectors, Cell Biolabs
Vectors, Plasmids and Libraries
Cell Biolabs' AAV Rep-Cap plasmids work with our AAV Helper Free Systems to produce recombinant AAV more safely and easily, without the need for a helper virus
Choose one of these plasmids if you already have the complete AAV Helper Free System and would like to package a different AAV serotype.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty