Would you like to log in again?
OxiSelect™ Ascorbic Acid Assay Kit (FRASC), Cell Biolabs
Supplier: Cell Biolabs
STA-860
102965-324EA 621.54 USD InStock
102965-324
OxiSelect™ Ascorbic Acid Assay Kit (FRASC), Cell Biolabs
Assays Cellular Assays
OxiSelect™ Ascorbic Acid Assay Kit is a quantitative assay for measuring ascorbic acid within various samples
The assay is based on the Ferric Reducing/Antioxidant Ascorbic Acid (FRASC) chemistry driven by the electron donating reducing power of antioxidants. The kit employs ascorbate oxidase, which allows the user to differentiate the ascorbic acid content from other antioxidants present within their sample. Ascorbic acid levels in a sample are determined by measuring the difference in optical density between two sample wells, one with and one without the enzyme.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty