Would you like to log in again?
TC Ryder Needle Holder, Serrated Tungsten Carbide, OR Grade, Sklar®
Supplier: Sklar
21-8023
89023-344EA 550.32 USD InStock
89023-344
TC Ryder Needle Holder, Serrated Tungsten Carbide, OR Grade, Sklar®
Forceps
Sklar's® TC Ryder Needle Holder is a ratcheted instrument that features narrow jaws and is commonly used with very small suture needles in cardiovascular, plastic, and neurosurgical procedures.
Tungsten carbide cross-serrated tips firmly grip the small needle and assist with keeping it from twisting during suturing.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty