Would you like to log in again?
TC New Orleans Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8082
89023-406EA 396.36 USD InStock
89023-406
TC New Orleans Needle Holder, OR Grade, Sklar®
Forceps
Sklar's® tungsten carbide New Orleans needle holder is a ratcheted instrument for holding and controling needles during suturing.
The tip of this instrument is short and features a single center groove which provides a secure hold on the needle
TC New Orleans and Censitrac Ready products are industry standard and constructed of stainless steel.
TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty