Would you like to log in again?
Meyerding Skin Hook, OR Grade, Sklar®
Supplier: Sklar
47-2462
89024-984EA 130.26 USD InStock
89024-984
Meyerding Skin Hook, OR Grade, Sklar®
Retractors
Sklar's® Meyerding Skin Hook is a double ended instrument featuring a right angle, flat, 15mm blade with a downward lip at one end and a sharp, thin 10mm hook at the other.
This retractor end is used for holding back muscle and tissue. The overall length is 61/4".
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty