Would you like to log in again?
Esmarch Plaster Shears, OR Grade, Sklar®
Supplier: Sklar
11-1380 11-1390
89025-470EA 288.43 USD InStock
89025-470 89025-472
Esmarch Plaster Shears, OR Grade, Sklar®
Scissors
Sklar® Esmarch Plaster Shears are non-ratcheted finger ring specialty scissors designed for use with plaster and fiberglass casting
Censitrac Ready products are industry standard and constructed of stainless steel.
These scissors should never be used on tissue. They are heavy shears with angled, standard blades.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty