Would you like to log in again?
Seutin Plaster Shears, OR Grade, Sklar®
Supplier: Sklar
11-1490
89025-474EA 596.34 USD InStock
89025-474
Seutin Plaster Shears, OR Grade, Sklar®
Scissors
Censitrac Ready OR Grade Seutin Plaster Shears are a single spring, plier-style instrument designed for use with plaster and fiberglass casting
They are heavy shears with blunt/blunt, angled blades.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty