Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TC Bruns Plaster Shears, OR Grade, Sklar®
Supplier: Sklar
16-2820
89025-730EA 897.47 USD InStock
89025-730
TC Bruns Plaster Shears, OR Grade, Sklar®
Scissors
Sklar® Bruns TC Plaster Shears are non-ratcheted finger ring specialty scissors designed for use with plaster and fiberglass casting
These scissors should never be used on tissue. They are heavy shears with one larger ring for the thumb. The lower blade has a blunted probe end so as not to cause injury to the patient while removing casting materials. It is also slightly longer than the standard blade. TC instruments are identified by gold handles. This product has slightly curved, smooth blades, blunt tips, and a 10cm stop peg in between the finger rings. It has overall length of 9-1/2 inches.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty