Would you like to log in again?
Joseph Button End Knife, OR Grade, Sklar
Supplier: Sklar
41-1305 41-1310 41-1300
10196-708EA 225.86 USD InStock
10196-708 10196-710 75817-564
Joseph Button End Knife, OR Grade, Sklar
Knives
Sklar's Joseph Button End Knife is an ENT/plastic surgery instrument most commonly used for dissecting and trimming nasal tissue
It is often used in rhinoplasty and septoplasty procedures. The small button tip of this instrument is used to separate nasal mucosa from the nasal cartilage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty