Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sklar's® Barraquer Wire Speculum, OR Grade, Sklar®
Supplier: Sklar
65-1740
10141-602EA 61.75 USD InStock
10141-602
Sklar's® Barraquer Wire Speculum, OR Grade, Sklar®
Specula
This is an ophthalmic instrument used in retracting the eye lid and holding it open during surgery.
  • Child size speculum with fenestrated blades
  • Relf-retaining retractor available
This speculum is a lightweight, spring style, wireform instrument, therefore minimal pressure is placed on the eye.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty