Would you like to log in again?
Spurling Intervertebral Disc Rongeur, OR Grade, Sklar®
Supplier: Sklar
42-2855
10196-944EA 871.69 USD InStock
10196-944
Spurling Intervertebral Disc Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Sklar's® Spurling IVD reongeur is used to debulk, manipulate or cut bone fragments and tissue from the spinal column.
  • Available in a greater variety of shaft lengths, up to 12"
Intervertebral disc rongeurs have cupped tips, available in differnt sizes, and either straight or angled up or down.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty