Would you like to log in again?
Surgi-OR™ TC Crile-Murray Needle Holder, Mid Grade, Sklar®
Supplier: Sklar
95-134
10184-342EA 133.85 USD InStock
10184-342
Surgi-OR™ TC Crile-Murray Needle Holder, Mid Grade, Sklar®
Forceps
The Surgi-OR™ TC Crile-Murray Needle Holder is used to hold and guide small to medium size needles and suture materials
The Crile-Murray Needle Holder is comparable to the Crile-Wood Needle Holder. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Hand-crafted in Pakistan, Surgi-OR™ TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with cross-serrated tips, and an overall length of 6 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty