Would you like to log in again?
Hartmann Mastoid Rongeur, OR Grade, Sklar®
Supplier: Sklar
67-5458
10198-400EA 480.64 USD InStock
10198-400
Hartmann Mastoid Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Sklar's® Hartmann Mastoid Rongeur is used for debulking in mastoidectomy procedures.
  • The Hartmann Mastoid Rongeur is used to remove portions of infected bone
  • This product is curved with smooth, blunt tips
Due to the use and effectiveness of antibiotics to treat ear infections, mastoidectomy procedures are not as common today as they were historically. Nonetheless, when antibiotic treatments are ineffective, infections of the middle ear may spread to the mastoid bone, thus making mastoidectomy necessary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty