Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Plain Wire Speculum, OR Grade, Sklar®
Supplier: Sklar
65-1127
10194-648EA 77.21 USD InStock
10194-648
Plain Wire Speculum, OR Grade, Sklar®
Specula
Sklar's® Plain Wire Eye Speculum is an opthalmic instrument used to separate and hold open the eyelids
The tension in this spring wire device allows the lightweight, open wire to gently retracts both the top and bottom eyelids without excessive pressure on the globe. The choice of eye speculum is typically dependent upon the anatomical nature of the patient's eyes. This product is an adult speculum with 15×7 mm open blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty