Would you like to log in again?
Harrington-Mixter Clamp, OR Grade, Sklar®
Supplier: Sklar
55-2912 55-3012
10197-592EA 440.61 USD InStock
10197-592 10197-590
Harrington-Mixter Clamp, OR Grade, Sklar®
Forceps
Sklar® Mixter Right Angle Forceps are ratcheted, finger ring forceps commonly used for working in obscured surgical sites
They are most frequently used for clamping, dissection, or grasping tissue. Mixter Right Angle Forceps are available in multiple lengths and have serrations the entire length of the jaw.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty