Would you like to log in again?
Doyen Jansen Mouth Gag, Pediatric, OR Grade, Sklar®
Supplier: Sklar
22-8945
10195-238EA 708.72 USD InStock
10195-238
Doyen Jansen Mouth Gag, Pediatric, OR Grade, Sklar®
Spatulas
Sklar's® Doyen-Jansen pediatric mouth gag, which may also be known as a Molt mouth gag, is used to retain the mouth opening during oral and ENT procedures.
It is a finger ring instrument that features a Tru-grip cam ratchet locking system. The blades of this instrument are 1" long and ¹/₂" wide. It also features cushions on the blades for the comfort of the patient.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty