Would you like to log in again?
Rap Activation Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-406-1 STA-406-2
102965-130EA 855.38 USD InStock
102965-130 102965-134
Rap Activation Assay Kits, Cell Biolabs
Assays Protein Assays
Cell Biolabs' Rap Activation Assays use visible agarose beads to selectively precipitate the active form of Rap
The precipitated small GTPase is then detected by Western blot using an Rap1- or Rap2-specific antibody included in the kit. The assays use a safe, non-radioactive format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty