Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Doyen Jansen Mouth Gag, Adult, OR Grade, Sklar®
Supplier: Sklar
75-1025
10198-628EA 910.48 USD InStock
10198-628
Doyen Jansen Mouth Gag, Adult, OR Grade, Sklar®
Spatulas
Sklar's® Doyen-Jansen adult mouth gag, which may also be known as a Molt mouth gag, is used to retain the mouth opening during oral and ENT procedures.
It is a finger ring instrument that features a Tru-grip cam ratchet locking system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty