Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ROCK Activity Immunoblot Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-415
102965-150EA 670.77 USD InStock
102965-150
ROCK Activity Immunoblot Kit, Cell Biolabs
Blotting Kits
Cell Biolabs' ROCK Activity Immunoblot Kit uses a safe, non-radioactive format to measure the level of active Rho Kinase in cell or tissue lysates
Rho-associated Kinase (ROCK) mediates Rho signaling and reorganizes the actin cytoskeleton by phosphorylation of several substrates that contribute to the assembly of actin filaments and contractility. This assay uses a safe, non-radioactive format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty