Would you like to log in again?
96-Well Ras Activation ELISA Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-441 STA-440
102965-164EA 1033.85 USD InStock
102965-164 102965-166
96-Well Ras Activation ELISA Kits, Cell Biolabs
Assays ELISAs
Cell Biolabs' 96-Well Ras Activation ELISA Kits use a standard 96-well plate format to quantify the active form of Ras using Raf1 RBD
Kits are available with either colorimetric or chemiluminescent detection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty