Would you like to log in again?
Alcian Blue Chondrogen Stain Kit, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0051
102970-836EA 212.92 USD InStock
102970-836
Alcian Blue Chondrogen Stain Kit, Lifeline Cell Technology
Stains Histology Stains
Lifeline® Alcian Blue Staining Kit is used for staining sulfated proteoglycans deposited by chondrocytes.
  • Fixative solution
  • Sucrose stabilizer solution
  • Alcian Blue stain solution
  • Acetic acid wash solution
Lifeline® Alcian Blue Staining Kit contains: fixative solution, sucrose stabilizer solution, Alcian Blue stain solution and acetic acid wash solution for staining sulfated proteoglycans deposited by chondrocytes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty