Would you like to log in again?
Oil Red O Lipid Staining Kit, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0052
102970-838EA 212.92 USD InStock
102970-838
Oil Red O Lipid Staining Kit, Lifeline Cell Technology
Stains Histology Stains
Lifeline® Oil Red O Staining Kit is used for staining lipid vesicles in adipocytes.
  • Fixative solution
  • Dehydration solution
  • Oil Red O stain solution
  • Stain differential solution
Lifeline® Oil Red O Staining Kit contains: fixative solution, dehydration solution, Oil Red O stain solution and stain differential solution for staining lipid vesicles in adipocytes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty