Would you like to log in again?
FibroLife® Serum-Free Fibroblast Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0001 LS-1010 LM-0001
102970-894EA 176.31 USD InStock
102970-894 102970-940 102970-758
FibroLife® Serum-Free Fibroblast Medium, Lifeline Cell Technology
Cell Culture Supplements and Additives
FibroLife® Serum-Free Medium Complete Kit is optimized for the serum-free culture of fibroblasts
  • Contains no phenol red or antimicrobials to affect cell performance.
  • Optimized for the serum-free culture of fibroblasts
  • Serum-free environment supports the growth of these cells
  • Maintains excellent cell morphology
The Complete Kit includes FibroLife® Basal Medium and FibroLife® Serum-Free LifeFactors® Kit (containing the associated supplements and growth factors) to allow you to prepare fresh medium. FibroLife® contains no phenol red or antimicrobials to affect cell performance.
FibroLife® Serum-Free LifeFactors® Kit includes: HLL Supplement (1.25 mL), rh FGF basic (0.5 mL), rh EGF/TGF β 1 (0.5 mL), L-Glutamine (18.75 mL), Hydrocortisone Hemisuccinate (0.5 mL), Ascorbic Acid (0.5 mL), rh Insulin (0.5 mL), Gentamicin and Amphotericin B (0.5 mL).
FibroLife® Complete Kits contain FibroLife® Basal Medium and the associated LifeFactors® Kit. FibroLife® Basal Medium requires the associated LifeFactors®.
FibroLife® Serum-Free supports the growth of these cells in a serum-free environment at rates equal to or greater than media supplemented with 2-10% FBS while maintaining excellent cell morphology.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty