Would you like to log in again?
Phosphatidylcholine Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-600
102965-204EA 578.46 USD InStock
102965-204
Phosphatidylcholine Assay Kit, Cell Biolabs
Assays Enzyme Assays
Cell Biolabs’ Phosphatidylcholine Assay Kit measures the phosphatidylcholine present within serum, plasma, or tissue samples
Samples are compared to a known concentration of phosphatidylcholine standard within the 96-well microtiter plate format.  Samples and standards are incubated for 60 minutes and then read with a standard 96-well fluorometric plate reader. Detect phosphatidylcholine concentrations as low as 0.5 mg/dL.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty