Would you like to log in again?
Sphingomyelin Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-601
102965-206EA 578.46 USD InStock
102965-206
Sphingomyelin Assay Kit, Cell Biolabs
Assays Enzyme Assays
Cell Biolabs’ Sphingomyelin Assay Kit is a simple fluorometric assay that measures the amount of sphingomyelin present in plasma or serum, tissue homogenates, or cell suspensionsin a 96-well microtiter plate format.  Each kit provides sufficient reagents to perform up to 96 assays, including blanks, sphingomyelin standards and unknown samples.  Sample sphingomyelin concentrations are determined by comparison with a known sphingomyelin standard.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty