Would you like to log in again?
MammaryLife™ Mammary Epithelial Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1088 LM-0041 LL-0061
102970-874EA 136.77 USD InStock
102970-874 102970-906 102970-790
MammaryLife™ Mammary Epithelial Medium, Lifeline Cell Technology
Cell Culture Media
Lifeline® MammaryLife™ Mammary Epithelial Medium is optimized for the culture of human mammary epithelial cells.
  • Supports human mammary epithelial cells
  • No phenol red
  • No antimicrobials
  • Guaranteed proliferation and function of cells
Lifeline® MammaryLife™ Mammary Epithelial Medium is optimized for the culture of human mammary epithelial cells. MammaryLife™ supports the growth of these cells over a period of at least 15 population doublings. MammaryLife™ contains no antimicrobials and no phenol red.
The MammaryLife™ Mammary Epithelial Medium Complete Kit includes:  MammaryLife™ Basal Medium and MammaryLife™ LifeFactors® Kit. MammaryLife™ basal medium requires the appropriate LifeFactors® to promote guaranteed proliferation and function of cells. MammaryLife™ LifeFactors® Kit includes: rh Insulin LifeFactor 0.5 mL, L-Glutamine LifeFactor 15 mL, Epinephrine LifeFactor 0.5 mL, Apo-Transferrin LifeFactor 0.5 mL, rh TGF-α LifeFactor 0.5 mL, Extract P™ LifeFactor 2 mL, Hydrocortisone Hemisuccinate LifeFactor 0.5 mL, Gentamicin and Amphotericin B 0.5 mL.
Ordering information: MammaryLife™ Medium Complete Kit includes MammaryLife™ Basal Medium and MammaryLife™ LifeFactors® Kit. MammaryLife™ Basal Medium requires the associated LifeFactors®.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty