Would you like to log in again?
Human Pulmonary Artery Endothelial Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0055
102970-704EA 1060 USD InStock
102970-704
Human Pulmonary Artery Endothelial Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Pulmonary Artery Endothelial Cells are isolated to ensure that they remain unexposed to recombinant human VEGF.
  • Isolated in VascuLife® EnGS Medium
  • No exposure to recombinant human VEGF
  • May be cultured with or without VEGF
Lifeline® Normal Human Pulmonary Artery Endothelial Cells (HPAEC) can be grown in low serum (2%) medium without phenol red or antimicrobials when cultured in VascuLife® medium
Lifeline® pulmonary artery endothelial cells are isolated in VascuLife® EnGS Medium to ensure that they remain unexposed to recombinant human VEGF.
Lifeline® pulmonary artery endothelial cells may be cultured with or without VEGF.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty