Would you like to log in again?
Ascorbic Acid LifeFactor, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1005
102970-980EA 69.85 USD InStock
102970-980
Ascorbic Acid LifeFactor, Lifeline Cell Technology
Cell Culture Supplements and Additives
Ascorbic Acid LifeFactor is used with human fibroblasts, cells and melanocytes, and basal medium.
  • Used with human endothelial cells
  • Used with human smooth muscle cells
  • Used with human fibroblasts
  • Used with human and neonatal melanocytes
  • Used with basal medium
Ascorbic Acid LifeFactor may be used with these Lifeline® products: a) FibroLife® basal medium and human fibroblasts, b) VascuLife® basal medium and human smooth muscle cells, c) VascuLife® basal medium and human endothelial cells, d) DermaLife basal medium and human melanocytes (neonatal, adult).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty