Would you like to log in again?
Pulmonary Artery Smooth Muscle Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0056
102970-706EA 1060 USD InStock
102970-706
Pulmonary Artery Smooth Muscle Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Pulmonary Artery Smooth Muscle Cells are quality tested and demonstrate optimal low serum growth.
  • Quality tested in VascuLife® SMC Medium
  • Demonstrate optimal low serum growth
  • No exposure to antimicrobials or phenol red
Lifeline® Normal Human Pulmonary Artery Smooth Muscle Cells are quality tested in VascuLife® SMC Medium and demonstrate optimal low serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lifeline® Human Pulmonary Artery Smooth Muscle Cells are not exposed to antimicrobials or phenol red.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty