Would you like to log in again?
L-Type Triglyceride M Assay, Wako
Supplier: Wako Diagnostics
992-02892 464-01601 992-02892 990-02991 994-02891 990-02991 464-01601 998-02992 998-02992 994-02891
10752-446PK 298.47 USD InStock
CA10752-444 10752-446 10752-444 CA10752-442 CA10752-440 10752-442 10752-440 CA10752-448 CA10752-446 10752-448
L-Type Triglyceride M Assay, Wako
Assays Enzyme Assays
L-Type triglyceride M is an in vitro assay for the quantification determination of triglyceride in serum or plasma.
  • Designed for use with Wako’s assays
  • Determine points of reference
  • Enzymatic method
The Wako Multi-Calibrator Lipids is designed to be used with Wako’s assays to determine the points of reference that are used in the determination of Triglycerides in human serum.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty