Would you like to log in again?
L-alanyl-L-glutamine, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1053
102970-752EA 67.54 USD InStock
102970-752
L-alanyl-L-glutamine, Lifeline Cell Technology
Cell Culture Supplements and Additives
L-alanyl-L-glutamine may be used with basal medium and stem cells.
  • Used with human mesenchymal stem cells
  • May be used as a stable substitute for L-glutamine
  • Minimal ammonia production
L-alanyl-L-glutamine may be used with Lifeline® StemLife MSC Basal Medium and human mesenchymal stem cells, including Wharton's Jelly and Adult. L-alanyl-L-glutamine may be used as a very stable substitute for L-glutamine, with the advantage of minimal ammonia production compared to L-glutamine in cell culture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty