Would you like to log in again?
Legionella GVPC Agar, bioMerieux
Supplier: bioMérieux
43032 43032
95060-808CS 372.54 USD InStock
95060-808 CA95060-808
Legionella GVPC Agar, bioMerieux
Media for Microbiology Dehydrated Media for Microbiology
Legionella GVPC Agar is a selective isolation of Legionella.
  • Selective isolation of Legionella
Selective isolation of Legionella.
Incubation: At least 10 days at 34-38°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty