Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HEPES, 1.0M buffer soln., pH 6.5, sterile-filtered

Supplier: Thermo Scientific
J60111-AK
AAJ60111-AKEA 269.73 USD InStock
AAJ60111-AK
HEPES, 1.0M buffer soln., pH 6.5, sterile-filtered
Buffers Biological Buffers
HEPES, 1.0M buffer soln., pH 6.5, sterile-filtered

CAS: 7365-45-9
Form: Liquid
Color: Clear, colorless
MDL Number: MFCD00006158
Storage Information: Keep Cold
TSCA: Yes

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty