Would you like to log in again?
Mayo-Adams Retractor, OR Grade, Sklar®
Supplier: Sklar
60-6467
10197-824EA 936.68 USD InStock
10197-824
Mayo-Adams Retractor, OR Grade, Sklar®
Retractors
Sklar's® Mayo-Adams Retractor is a handheld self-retaining instrument that features a Tru-Grip ratchet
This retractor is used in a variety of procedures to hold back tissue and expose the surgical site.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty