Would you like to log in again?
HumaCell, Access Cell Culture
Supplier: Access Cell Culture
HumaCell
525-100HI 525
103039-158EA 135.38 USD InStock
103039-158 103012-080
HumaCell, Access Cell Culture
Sera (Serum)
HumaCell is human serum that is not gender specific and it is not blood type specific
  • Mixed gender, mixed blood type human serum
  • Ideal for customers using FBS to cultivate human cell lines
  • Ideal, but not limited to the cultivating of: dermal fibroblasts, bone marrow, Mesenchymal Stem Cells and PBMC’s
  • Can be used for therapeutic use
HumaCell is human serum that is not gender specific and not blood type specific
Our heat Inactivation protocol involves bringing the water bath temperature to 56° C. This temperature is maintained for 30 minutes.
It is collected from donors meeting the health requirements established by 21CFR 640 subpart G via plasmapharesis. These individual units are converted into serum and then pooled, filtered, bottled and cell culture tested for mycoplasma, endotoxin, and USP sterility. Each individual unit is tested and found negative for all required viral markers via FDA-approved methods.
Certifications: UL Listed. CE compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty