Would you like to log in again?
Rongeur No. 5S Side Cut, OR Grade, Sklar®
Supplier: Sklar
40-3196
10195-632EA 497.99 USD InStock
10195-632
Rongeur No. 5S Side Cut, OR Grade, Sklar®
Bone Cutters and Rongeurs
Side cutting Cleveland rongeur are orthopedic heavy duty forceps for gouging out bone or removing tough tissue. This is the single action spring plier handle type, similar to the Dean, Luer and Olivcrona.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty