Would you like to log in again?
Cooley Anastomosis Clamp, OR Grade, Sklar®
Supplier: Sklar
52-6960
10197-096EA 645.88 USD InStock
10197-096
Cooley Anastomosis Clamp, OR Grade, Sklar®
Forceps
Sklar's® Cooley Atraumatic Anastomosis Clamp is a ratcheted, finger ring instrument used to control blood flow from vessels during anastomosis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty