Would you like to log in again?
Tube Occluding Clamp, OR Grade, Sklar
Supplier: Sklar
17-3997 17-3995 17-3996
82030-026EA 215.28 USD InStock
82030-026 82030-024 82030-022
Tube Occluding Clamp, OR Grade, Sklar
Forceps
Sklar® Presbyterian Tube Occluding Forceps are a ratcheted, instrument used to control moderate to heavy flow of fluids or blood. They are a heavyweight instrument and can be used with heavy tubing or vessels.
The jaws and tips of this instrument are smooth so as not to cause damage to the vessel being occluded
Censitrac Ready products are industry standard and constructed of stainless steel.
These forceps should not be used on patient tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty