Would you like to log in again?
Reusable Steel Knives, Epredia™
Supplier: Epredia
Epredia™
152100 152090 152080 152270 152060 152070 152010 152020 152050
89235-246EA 705.55 USD InStock
89235-246 89235-258 89235-244 89235-256 89235-260 89235-250 89235-270 89235-254 89235-252
Reusable Steel Knives, Epredia™
Knives
Use Epredia™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Permanent knives can be resharpened on an appropriate knife sharpener for extended use
C-profile knives are used for standard paraffin and cryo techniques. D-profile knives are used for sectioning of harder materials that require greater knife stability. Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin.
Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty