Would you like to log in again?
Shandon™ Glyo-Fixx, Thermo Scientific
Supplier: Thermo Scientific
Glyo-Fixx™
6764265 6764265
89369-334EA 153.89 USD InStock
CA89369-334 89369-334
Shandon™ Glyo-Fixx, Thermo Scientific
Stains Microscopy and staining reagents
Obtain superior results to neutral buffered formalin with this noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Available in Ready-To-Use and Concentrate solution
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Presents fine detail in specimen structure
Allows special stain dyes, including silver procedures, to be sharp without background staining.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty