Would you like to log in again?
Lab Vision™ UltraVision™ DAB Away™ Thermo Scientific
Supplier: Richard-Allan Scientific
Lab Vision™ UltraVision™ DAB Away™
TA-250-DA
10129-596EA 538.65 USD InStock
10129-596
Lab Vision™ UltraVision™ DAB Away™ Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
Remove stains and diaminobenzidine precipitate from your autostainer using UltraVision DAB Away.
Apply to stained glassware, staining pads, and more.
Store in room temperature. Protect from light.
Ordering information: Includes: DAB Away Reagent 1 (125 mL), DAB Away Reagent 2 (125 mL), DAB Away Decolorizer (250 mL).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty