Would you like to log in again?
UltraVision™ Quanto Mouse on Mouse, Thermo Scientific
Supplier: Epredia
TL-060-QHDM
89212-328EA 2895.54 USD InStock
89212-328
UltraVision™ Quanto Mouse on Mouse, Thermo Scientific
Protein Linking and Labeling Systems
UltraVision Quanto Mouse on Mouse HRP IHC detection system makes optimal results routine with mouse antibodies on mouse tissue.
UltraVision Quanto Makes Optimal Results Routine by providing: Strong StainingReduced Mouse on Mouse Background
Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Thermo Scientific UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely.
Simple Biotin Free Polymer Procedure. For use with a DAB or other HRP Chromogen. For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty