Would you like to log in again?
AimStrip® Hemoglobin Test, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip®
78250 78101 78102 78200 78103
10024-814EA 385.59 USD InStock
10024-814 10024-816 10024-820 10024-812 10024-822
AimStrip® Hemoglobin Test, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Analysers
The AimStrip® Hemoglobin Test is designed for the quantitative determination of hemoglobin in non-anticoagulated capillary whole blood or anticoagulated venous whole blood in EDTA
Results are provided in 15 seconds. Hb measurements range from 5.6 to 23.5g/dL. The handheld meter uses batteries or an optional AC adapter, stores up to 1,000 tests, and features an automatic shutoff after 8 minutes of non-use. Meter is supplied with a three-year warranty.
Estimation of hematocrit is provided for normal hemoglobin values. This device has not been evaluated for pediatric subjects.
Certifications: CLIA waived.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty