Would you like to log in again?
AIM™ ASO Test, Germaine Laboratories
Supplier: Germaine Laboratories
AIM™
31050
10024-414BX 62.23 USD InStock
10024-414
AIM™ ASO Test, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
This latex agglutination test is designed for the qualitative and semi-quantitative determination of Antistreptolysin-O (ASO) in non-diluted serum
This test is recommended for professional use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty