Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Drum Pallet Truck, Wesco

Supplier: Wesco
273250
89093-020EA 3023.16 USD InStock
89093-020
Drum Pallet Truck, Wesco
Carts
Hydraulic pallet truck designed to lift and move 55 gallon steel drums.

  • Lowered height 10"
  • Raised height 14.5"
  • Lifts drum 4" off ground
  • Fork foot print 28.5x26"

8x2" nylon steering wheels; 1x3" nylon load wheels.

Ordering information: Manufacturer's one-year warranty on pump.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty