Would you like to log in again?
Airway Trauma Response System™, Emergency Medical International
Supplier: Emergency Medical International
879
10853-014KT 372.96 USD InStock
10853-014
Airway Trauma Response System™, Emergency Medical International
First Aid Supplies First Aid Kits
EMI’s Airway Trauma Response System™ provides a single source for carrying oxygen, airways, and intubation and trauma equipment.
  • A drawstring oxygen carrying bag with web carrying handles is also included
  • Pack has a star of life reflective emblem on front and yellow reflective trim on the front and the sides
  • Pack measures 58.4L×30.5W×22.9H cm (23×12×9")
Lightweight, durable, rugged, and compact, it is made of 1000 denier nylon. All of the pack walls are encircled with a layer of 0.25" rigid foam rubber, and then covered with an additional layer of softer foam for extra protection. A detachable laryngoscope/airway panel fits securely by velcro into the lid of the pack.
The pack also has five clear removable field packs (two large and three small) and two additional zippered mesh pockets. There is also extra storage in the large front outside zippered compartment. C or 'D' tanks are secured by two velcro straps and buckles.
Ordering information: Includes drawstring oxygen carrying bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty