Would you like to log in again?
Human Recombinant Aldose Reductase (from E. coli)
Supplier: BioVision
AB275926-100UG
10834-242EA 559.09 USD InStock
10834-242
Human Recombinant Aldose Reductase (from E. coli)
Enzymes
≥95% pure active human Recombinant Aldose Reductase
Aldose reductase (AKR1B1) is a member of the aldo-keto reductase (AKR) superfamily and catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols. This protein is implicated in the development of diabetic complications by catalyzing the reduction of glucose to sorbitol. Recombinant Aldose reductase (AKR1B) protein was expressed in E.coli and purified by using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty