Would you like to log in again?
Human Native alpha-1-Acid Glycoprotein (from Plasma)
Supplier: BioVision
AB285894-10MG AB285894-2MG
10835-656EA 459.8 USD InStock
10835-656 10835-654
Human Native alpha-1-Acid Glycoprotein (from Plasma)
Proteins and Peptides
≥95% pure native alpha 1 acid glycoprotein isolated from human plasma
Found in human plasma in concentrations of 55-14 mg per 1 ml. Classical standard glycoprotein for studies on the structure of the oligosaccharide units. Carbohydrate content is 4%. Its biological significance is unknown although it can bind progesterone 15 times as strongly as albumin. Sialic-acid-deficient alpha-1-AG has an affinity for vitamin B-12. Clinically, alpha 1 acid glycoprotein is an acute-phase reactant that together with haptoglobin is an indicator of acute inflammation. The alpha 1 acid glycoprotein: haptoglobin ratio is useful in studies of bone marrow disorders, hemolytic processes and metastases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty